Foxp3 / Transcription Factor Staining Buffer Kit, Tonbo Biosciences
Lieferant: Tonbo Biosciences
Tonbo Biosciences Foxp3 / Transcription Factor Staining Buffer Kit contains specially formulated buffers and solutions for optimal resolution and low background in your analysis of nuclear antigens by flow cytometry
Foxp3 / Transcription Factor Fix/Perm Concentrate (4X) 50 ml. A concentrated solution which, when diluted with Foxp3 / Transcription Factor Fix/Perm Diluent (1X) provides best results in protocols for intranuclear staining of transcription factors using fluorescently conjugated antibodies.
Foxp3 / Transcription Factor Fix/Perm Diluent (1X) 160 ml. Intended for use as a diluent for Foxp3 / Transcription Factor Fix/Perm Concentrate (4X).
Flow Cytometry Perm Buffer (10X) 150 ml. Provided as a concentrate which, when diluted with distilled water to a 1X solution, provides best results in intracellular staining protocols for cytokines and other cytoplasmic antigens, by maintaining membrane permeabilization throughout staining and wash steps.
This complete kit contains three components for use in these staining protocols for detection of nuclear antigens such as Foxp3 and ROR gamma.
Tested applications: Flow Cytometry
Reactivity: Mouse, Human
Storage temperature: Store Undiluted at 2 to 8 °C; Protect from Light; Do Not Freeze.
Bestellinformation: Kit includes (1) Foxp3 / Transcription Factor Fix/Perm Concentrate (4X) 50 ml. A concentrated solution which, when diluted with Foxp3 / Transcription Factor Fix/Perm Diluent (1X) provides best results in protocols for intranuclear staining of transcription factors using fluorescently conjugated antibodies; (2) Foxp3 / Transcription Factor Fix/Perm Diluent (1X) 160 ml. Intended for use as a diluent for Foxp3 / Transcription Factor Fix/Perm Concentrate (4X); (3) Flow Cytometry Perm Buffer (10X) 150 ml. Provided as a concentrate which, when diluted with distilled water to a 1X solution provides best results in intracellular staining protocols for cytokines and other cytoplasmic antigens, by maintaining membrane permeabilization throughout staining and wash steps.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...