Surfactant, RapiGest™ SF
Lieferant: WATERS
186001861
WATE186001861EA
375
EUR
WATE186001861
Surfactant, RapiGest™ SF
Protein-Extraktion
Proteinextraktions-Reagenzien
RapiGest™ SF is an anionic surfactant for peptide digestion. Supplied as five 1 mg vials for 1 to 50 digestions. The digestion speed and more peptide recovery is enhanced by using trypsin or other enzymes.