Anti-CD3E Rat Monoclonal Antibody [clone: 17A2]

Lieferant: Tonbo Biosciences
40-0032-U500 40-0032-U100
40-0032-U500EA 142 EUR
40-0032-U500 40-0032-U100
Anti-CD3E Rat Monoclonal Antibody [clone: 17A2]
Antikörper
The 17A2 antibody reacts with the mouse CD3 complex, comprised of CD3 epsilon, CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells.

The 17A2 antibody is a widely used phenotypic marker for mouse T cells. In addition, as the CD3e chain within the TCR complex contains intracellular signaling domains, binding of 17A2 antibody to CD3 can induce cell activation (use format suitable for functional assays). A recent publication of the crystal structure of a murine CD3e-mitogenic antibody complex provides further insight into the action of commonly used agonist antibodies (Fernandes, R.A. et al. 2012. Journal of Biological Chemistry. 287: 13324-13335).

Type: Primary
Antigen: CD3E (CD3e molecule, epsilon (CD3-TCR complex))
Clonality: monoclonal
Clone: 17A2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR