Anti-TCRB Armenian Hamster Monoclonal Antibody (PE) [clone: H57-597]
Lieferant: Tonbo Biosciences
50-5961-U025 50-5961-U100
50-5961-U025EA 45.8 EUR
50-5961-U025 50-5961-U100
Anti-TCRB Armenian Hamster Monoclonal Antibody (PE) [clone: H57-597]
Antikörper
The H57-597 antibody is specific for the beta chain of the mouse T cell Receptor (TCR). This cell surface protein combines with a second protein chain (alpha chain) to form the alpha-beta TCR that is expressed by NK1.1+ thymocytes, NKT cells, and the majority of peripheral T cells. A small number of T cells may express an alternative heteromer of gamma/delta protein chains, known as the g/d TCR. These receptors participate in a complex with CD3, and with the co-receptors CD4 or CD8, to recognize and respond to antigens bound to MHC molecules on antigen-presenting cells. Such interactionsMore Product Information