Anti-HSPD1 Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ADISPS875D ADISPS875F
ENZOADISPS875DEA 225 EUR
ENZOADISPS875D ENZOADISPS875F
Anti-HSPD1 Rabbit Polyclonal Antibody
Antikörper
GroEL and GroES are E. coli chaperonins, which are homologs of mitochondrial chaperonins Hsp60 and Hsp10. GroEL is a double toriodal assembly of 14 identical subunits which form two heptameric rings stacked back-to-back, with a cavity at each end. GroEL and its co-chaperonin GroES facilitate protein folding with an ATP-dependent mechanism.

  • Dilution range: Western blot (1:1,000, colorimetric)
  • Detects a band of ~60 kDa by western blot

Type: Primary
Antigen: GroEL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: E.coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR