Anti-LAMP2 Rat Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ADIVAMEN003D ADIVAMEN003F
ENZOADIVAMEN003DEA 288 EUR
ENZOADIVAMEN003D ENZOADIVAMEN003F
Anti-LAMP2 Rat Monoclonal Antibody
Antikörper
Lysosome-associated membrane proteins (LAMP1 and LAMP2) are type I integral membrane protein that are transported from the trans-Golgi network to endosomes and then lysosomes. Following activation of platelets, T-cells, neutrophils, or endothelium, LAMP1 and LAMP2 are expressed on the cell surface. Cell surface LAMP1 and LAMP2 have been shown to promote adhesion of human peripheral blood mononuclear cells (PBMC) to vascular endothelium, and loss of LAMP2 expression is associated with impaired autophagy observed in Danons disease.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes mouse and rabbit LAMP2.

Type: Primary
Antigen: LAMP2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR