Anti-SAP97 Mouse Monoclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ADIVAMPS005D ADIVAMPS005F
ENZOADIVAMPS005DEA 275 EUR
ENZOADIVAMPS005D ENZOADIVAMPS005F
Anti-SAP97 Mouse Monoclonal Antibody
Antikörper
Synapse-associated proteins SAP97 and SAP102 display extensive sequence homology to the PSD 95 family of proteins that facilitate ion channel clustering at synaptic terminals. In addition to the CNS, SAP97 is also detected at the basal lateral membrane between a variety of epithelial cells. SAP102 associates with NMDA receptors through the C-terminal NR2B subunit of the receptor. The N-terminal SAP97 PDZ domain mediates its interaction with the synaptic ras-GTPase activating protein SynGAP.
  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot, EM
Recognizes human, mouse, rat, and dog SAP97. Detects bands of ~97kDa and ~140kDa by Western blot.
Type: Primary
Antigen: SAP97
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human