Anti-MRP4 Rat Monoclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX801039C100 ALX801039C200
ENZOALX801039C100EA 438 EUR
ENZOALX801039C100 ENZOALX801039C200
Anti-MRP4 Rat Monoclonal Antibody
Antikörper
Host: Rat, Isotype: IgG2a
  • For Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes human and mouse MRP4.
Type: Primary
Antigen: MRP4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human