Anti-CD163 Rabbit Polyclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX810213R100 ALX810213R500
ENZOALX810213R100EA 255 EUR
ENZOALX810213R100 ENZOALX810213R500
Anti-CD163 Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit, Isotype: IgG
  • For Immunohistochemistry
Recognizes human CD163.
Type: Primary
Antigen: CD163
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human