Anti-PARP Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
BMLSA2530025 BMLSA2530100
ENZOBMLSA2530025EA 160 EUR
ENZOBMLSA2530025 ENZOBMLSA2530100
Anti-PARP Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human, mouse, rat and bovine PARP. Detects a band of ~116kDa by Western blot.

Type: Primary
Antigen: PARP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR