Anti-GRP78 Rabbit Polyclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ADISPA7680050 ADISPA7680200
ENZOADISPA7680050EA 184 EUR
ENZOADISPA7680050 ENZOADISPA7680200
Anti-GRP78 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Grp78
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Monkey, Hamster, Guinea pig, Fungus, Dog, Bovine