Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (FITC)

Lieferant: STEMCELL Technologies
100-0285 100-0286
STMC100-0285EA 129 EUR
STMC100-0285 STMC100-0286
Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (FITC)
Antikörper
Mouse monoclonal IgG2a, antibody against human CD3, FITC-conjugated.

  • T cells

The OKT3 antibody reacts with an extracellular conformational epitope on the ~20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ~95% of mature T cells and NKT cells, and variably on thymocytes. A majority of T cell neoplasms also express CD3. CD3 is a dimer assembled from combinations of CD3γ, δ, ε, η, and ζ subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily. CD3 associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of T cells and in regulating T cell activation and the expression of the TCR on the cell surface. Mutations in CD3 have been associated with disorders of the immune system, including severe combined immunodeficiency (SCID). The OKT3 antibody recognizes an epitope that is expressed when the CD3ε subunit associates with either CD3γ or CD3δ, and binding of the antibody can induce T cell activation. Because of its immunosuppressive properties in vivo, the OKT3 antibody has proven useful in treating allograft rejection. Clone OKT3 is widely used for phenotyping human T cells, and it has been observed to block the binding of anti-CD3 antibody clones SK7 and UCHT1.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR